Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Prepared Lysine Iron Agar (LIA) Media Slants

Catalog No. BB20953 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BB20953 100 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BB20953 Supplier BD Supplier No. 220953
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For differentiation of enteric organisms based on their ability to decarboxylate or deaminate lysine and to form hydrogen sulfide

  • Available as slants (K tubes)

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, AOAC, BAM, CCAM, CMPH2, COMPF, MCM9, SMD, SMWW, USDA
Color Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Lysine Iron Agar
Formulation Pancreatic digest of Gelatin 5g/L, Yeast extract 3g/L, Dextrose 1g/L, L-Lysine 10g/L, Ferric Ammonium Citrate 0.5g/L, Sodium Thiosulfate 0.04g/L, Bromcresol Purple 0.02g/L, Agar 13.5g/L
For Use With (Application) For differentiation of enteric organisms based on their ability to decarboxylate or deaminate Lysine and to form Hydrogen sulfide
Packaging Type Tube
Quantity 100 Pack

For in vitro diagnostic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.