Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Prepared Pour Tubed Media: Trypticase™ Soy Agar (Blood Agar Base) Deeps

Catalog No. BB21082 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BB21082 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BB21082 Supplier BD Supplier No. 221082
Add to Cart
Add to Cart

For the isolation and cultivation of fastidious and nonfastidious anaerobic and aerobic microorganisms

  • For cultures requiring prolonged incubation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP
Color Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Trypticase soy agar
Product Type Prepared Microbiology Tubes
Test Type Microbial Cultivation
Fill Volume 20 mL
Formulation Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 15g/L
For Use With (Application) Isolation and cultivation of non-fastidious and fastidious microorganisms
No. per Pack 10
Packaging Type Tube
Quantity 20 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.