Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Prepared Tubed Media: BHI Agar Slant w/Sheep Blood, w/Chloramphenicol, Gentamicin

Catalog No. L95756 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
ISO/DIS, 11014, BS12, CMPH2, MCM9
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
L95756 ISO/DIS, 11014, BS12, CMPH2, MCM9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L95756 Supplier BD Supplier No. 295756
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in qualitative procedures for the isolation and cultivation of pathogenic and nonpathogenic fungi from clinical and nonclinical specimens

  • For cultures requiring prolonged incubation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, BS12, CMPH2, MCM9
Color Red
Content And Storage 2°C to 8°C
Additive Chloramphenicol, Gentamicin
Detects Tubercle bacilli, Fungi
Form Liquid
Manufacturing Origin United States
Media Type BHI agar slant with Sheep blood
Product Type Agar Slants
Sterility Sterile
Target Organism Class Fungi
Test Type Microbial Cultivation
Formulation Brain heart infusion from (solids) 8g/L, Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Casein 16g/L, Dextrose 2g/L, Sodium Chloride 5g/L, Disodium Phosphate 2.5g/L, Agar 13.5g/L
For Use With (Application) Isolation and cultivation of pathogenic and nonpathogenic fungi
No. per Pack 100
Packaging Type Tube
Quantity 100/Pk.
Protein Source Animal Tissue, Casein, Sheep Blood
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.