Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Prepared Tubed Media: BHI Broth

Catalog No. L21813 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
5mL, Carton of 100 Size K Tubes
8mL, Carton of 100 Size K Tubes
Media Type:
Brain Heart Infusion Broth
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Media Type
L21813 5mL, Carton of 100 Size K Tubes Brain Heart Infusion Broth
BB20837 8mL, Carton of 100 Size K Tubes Brain Heart Infusion Broth
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. L21813 Supplier BD Supplier No. 221813
Add to Cart
Add to Cart
This item is not returnable. View return policy

General-purpose liquid media used in the cultivation of fastidious and nonfastidious microorganisms, including aerobic and anaerobic bacteria, from a variety of clinical and nonclinical materials

  • For cultures requiring prolonged incubation

TRUSTED_SUSTAINABILITY

Specifications

Broth Type BHI Broth
Certifications/Compliance ISO/DIS, 11014, AOAC, BAM, CCAM, CLSI, CMPH2, COMPF, EPA, ISO, MCM9, SMD, SMWW, USDA
Color Clear
Detects Tubercle Bacilli, Fungi, Fastidious Microorganisms, Non-fastidious Microorganisms
Form Liquid
Manufacturing Origin United States
Media Type Brain Heart Infusion Broth
Product Type Broth Tube
Sterility Sterile
Test Type Microbial Cultivation
Fill Volume 5 mL
Formulation Brain heart infusion from (solids) 6g/L, Peptic digest of Animal Tissue 6g/L, Sodium Chloride 5g/L, Dextrose 3g/L, Pancreatic digest of Gelatin 14.5g/L, Disodium Phosphate 2.5g/L
For Use With (Application) Cultivation of fastidious and non-fastidious microorganisms
Includes 100 K-tubes
No. per Pack 100
Packaging Type K Tube
Quantity 5mL, Carton of 100 Size K Tubes
Protein Source Animal Tissue, Gelatin
Manufacturing Quality ISO
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.