Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Prepared Tubed Media: BHI broth with 6.5% NaCl

Catalog No. L21785 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
0.5 mL Tubes, 10 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L21785 0.5 mL Tubes, 10 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L21785 Supplier BD Supplier No. 221785
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For cultivation of fastidious and nonfastidious microorganisms from a vriety of clinical and nonclinical materials

  • For cultures requiring prolonged incubation
  • Contains 6.5% sodium chloride

TRUSTED_SUSTAINABILITY

Specifications

Broth Type BHI Broth with 6.5% NaCl
Certifications/Compliance ISO/DIS, 11014, AOAC, BAM, CCAM, CLSI, CMPH2, COMPF, EPA, ISO, MCM9, SMD, SMWW, USDA
Color Beige
Concentration 6.5% NaCl
Content And Storage 2 to 25°C
Additive Sodium Chloride
Detects Microorganisms, Tubercle bacilli, Fungi
Form Liquid
Manufacturing Origin United States
Media Type Brain heart infusion broth
Sterility Sterile
Target Organism Class Fastidious Organisms
Formulation Brain heart infusion from (solids) 6g/L, Peptic digest of Animal Tissue 6g/L, Sodium Chloride 5g/L, Dextrose 3g/L, Pancreatic digest of Gelatin 14.5g/L, Disodium Phosphate 2.5g/L
For Use With (Application) General purpose liquid medium used in the cultivation of fastidious and non-fastidious microorganisms
Includes 10 Tubes
No. per Pack 10
Packaging Type Tube
Quantity 0.5 mL Tubes, 10 Pack
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.