Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Prepared Tubed Media: Bile Esculin Agar Slants

Catalog No. BB21410 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100/Pk.
For Use With (Application):
Isolation and cultivation of pathogenic and nonpathogenic fungi
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity For Use With (Application)
BB21410 100/Pk. Isolation and cultivation of pathogenic and nonpathogenic fungi
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BB21410 Supplier BD Supplier No. 221410
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used to differentiate enterococci and the Streptococcus bovis group from other streptococci

  • For cultures requiring prolonged incubation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, COMPF
Color Beige
Content And Storage 2°C to 8°C
Additive Ferric Citrate, Oxgall
Detects Enterococci, Streptococcus bovis Group
Form Solid
Format Solid, Agar
Manufacturing Origin United States
Media Type Bile Esculin Agar
Product Type Agar Slants
Sterility Sterile
Target Organism Class Bacteria
Test Type Microbial Differentiation
Formulation Pancreatic digest of Gelatin 5g/L, Beef extract 3g/L, Oxgall 20g/L, Ferric Citrate 0.5g/L, Esculin 1g/L, Agar 14g/L
For Use With (Application) Isolation and cultivation of pathogenic and nonpathogenic fungi
Incubation Conditions 35 ±2°C, Aerobic Atmosphere
No. per Pack 100
Packaging Type Tube
Quantity 100/Pk.
Protein Source Pancreatic Digest of Gelatin, Beef Extract
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.