Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Prepared Tubed Media: Bile Esculin Azide Enterococcosel™ Broth

Catalog No. L21383 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
CE, CMPH2, ISO, MCM9
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
L21383 CE, CMPH2, ISO, MCM9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L21383 Supplier BD Supplier No. 221383
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use in the differentiation of enterococci

  • For cultures requiring prolonged incubation

TRUSTED_SUSTAINABILITY

Specifications

Appearance Yellow Brown Liquid
Broth Type Bile Sculin Azide Enterococcosel Broth
Certifications/Compliance CE, CMPH2, ISO, MCM9
Color Yellow Brown
Content And Storage 2°C to 8°C
Detects Enterococci
Form Liquid
Format Liquid, Tube
Manufacturing Origin United States
Media Type Bile esculin azide Enterococcosel broth
Product Type Broth Tube
Sterility Sterile
Target Organism Class Non-fastidious Organisms
Test Type Microbial Differentiation
Formulation Pancreatic digest of Casein 17g/L, Peptic digest of Animal Tissue 3g/L, Yeast extract 5g/L, Oxgall 10g/L, Sodium Chloride 5g/L, Sodium Citrate 1g/L, Esculin 1g/L, Ferric Ammonium Citrate 0.5g/L, Sodium Azide 0.25g/L
For Use With (Application) Isolation and cultivation of non-fastidious and fastidious microorganisms
Incubation Time 8 Weeks or more
No. per Pack 10
Packaging Type Tube
Quantity 10/Pk.
Manufacturing Quality ISO
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.