Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Prepared Tubed Media: Blood Agar Slant

Catalog No. BB20830 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
CE, BS12, CMPH2, MCM9
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
BB20830 CE, BS12, CMPH2, MCM9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BB20830 Supplier BD Supplier No. 220830
Add to Cart
Add to Cart
This item is not returnable. View return policy

For cultivating and maintaining fastidious microorganisms

  • For cultures requiring prolonged incubation
  • Trypticase™ soy agar with defibrinated sheep blood slant

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BS12, CMPH2, MCM9
Color Red
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Blood agar
Formulation Pancreatic digest of Casein 14.5g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 14g/L, Growth Factors 1.5g/L, Sheep Blood (Defibrinated) 10%
For Use With (Application) Isolation and identification of microorganisms requiring prolonged incubation
No. per Pack 10
Packaging Type Tube
Quantity 10/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.