Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Prepared Tubed Media: Rappaport Vassiliadis Soy Salmonella Broth

Catalog No. 22349976 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-349-976 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-349-976 Supplier BD Supplier No. 215199
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For isolation and identification of microorganisms and for cultures requiring prolonged incubation.

  • Ready-to-use
  • Non-sterile
  • Green liquid

TRUSTED_SUSTAINABILITY

Specifications

Broth Type Rappaport-Vassiliadis Soy Salmonella Broth
Certifications/Compliance USP, ISO/DIS, 11014
Color Green
Content And Storage 2 to 25°C
Detects Salmonella spp., Tubercle bacilli, Fungi
Form Liquid
Format Liquid, Tube, Ready-to-use
Manufacturing Origin United States
Media Type Rappaport Vassiliadis Soy Salmonella broth
Product Type Prepared Microbiology Tubes
Sterility Sterile
Test Type Microbial Identification
Fill Volume 10 mL
For Use With (Application) Isolate and Cultivate Tubercle bacilli and Fungi
No. per Pack 10
Packaging Type Tube
Quantity 10/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.