Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Sabouraud Brain Heart Infusion Agar Prepared Media Slants

Catalog No. L97691 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
ISO/DIS, 11014, BS12, MCM9
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
L97691 ISO/DIS, 11014, BS12, MCM9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L97691 Supplier BD Supplier No. 297691
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used in qualitative procedures for isolation and cultivation of pathogenic fungi from clinical and nonclinical specimens

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, BS12, MCM9
Color Light Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Sabouraud Brain Heart Infusion Agar
Formulation Brain heart infusion from (solids) 4g/L, Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Casein 10.5g/L, Dextrose 21g/L, Sodium Chloride 2.5g/L, Disodium Phosphate 1.25g/L, Agar 15g/L
For Use With (Application) Used for qualitative procedures for isolation and cultivation of pathogenic fungi from clinical and nonclinical specimens
No. per Pack 100
Packaging Type Tube
Quantity 100 Pack
Show More Show Less

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.