Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Sabouraud Brain Heart Infusion Agar Slants, Chloramphenicol, Gentamicin

Catalog No. L97252 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
ISO/DIS, 11014, MCM9
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
L97252 ISO/DIS, 11014, MCM9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L97252 Supplier BD Supplier No. 297252
Add to Cart
Add to Cart

For selective isolation and cultivation of pathogenic fungi from clinical and nonclinical specimens

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, MCM9
Content And Storage 2°C to 8°C
Detects Pathogenic Fungi
Form Liquid
Format Liquid, Agar Slants, Tube
Manufacturing Origin United States
Media Type Sabouraud Brain Heart Infusion Agar
Product Type Agar Slants
Target Organism Class Fungi
Test Type Microbial Cultivation
Formulation Brain heart infusion from (solids) 4g/L, Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Casein 10.5g/L, Dextrose 21g/L, Sodium Chloride 2.5g/L, Disodium Phosphate 1.25g/L, Agar 15g/L, Chloramphenicol 0.05g/L, Gentamicin 0.05g/L
For Use With (Application) Qualitative procedures for the selective isolation and cultivation of pathogenic fungi from clinical and nonclinical specimens
No. per Pack 10
Packaging Type Tube
Quantity 10/Pk.
Show More Show Less

For in vitro diagnostic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.