Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Sabouraud Dextrose Agar Prepared Media Slants

Catalog No. BB21013 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BB21013 100 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BB21013 Supplier BD Supplier No. 221013
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used in qualitative procedures for cultivation of dermatophytes

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Cap Type Tight Cap
Certifications/Compliance CE, BAM, CCAM, CMPH2, COMPF, EP, JP, MCM9, USP
Color Beige
Content And Storage 2°C to 8°C
Detects Dermatophytes
Form Liquid
Format Agar Slant, Tube
Manufacturing Origin United States
Media Type Sabouraud Dextrose Agar
Product Type Agar Slants
Sterility Sterile
Target Organism Class Fungi
Test Type Microbial Cultivation
Fill Volume 10 mL
Formulation Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 40g/L, Agar 15g/L
For Use With (Application) Cultivation of dermatophytes
Includes Mycoflask, Tube with Tight Cap
No. per Pack 100
Packaging Type Tube with Tight Cap
Quantity 100 Pack
Protein Source Casein, Animal Tissue
Manufacturing Quality CE, BAM, CCAM, CMPH2, COMPF, EP, JP, MCM9, USP
Show More Show Less

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.