Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Sabouraud Dextrose Agar Prepared Media Slants with Chloramphenicol

Catalog No. L21825 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L21825 100 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L21825 Supplier BD Supplier No. 221825
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used in qualitative procedures for cultivation of dermatophytes

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, MCM9
Color Beige
Content And Storage 2°C to 8°C
Detects Fungi, Tubercle bacilli
Form Liquid
Manufacturing Origin United States
Media Type Sabouraud Dextrose Agar
Product Type Agar Slants
Sterility Sterile
Target Organism Class Fungi
Formulation Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 40g/L, Agar 15g/L
For Use With (Application) Cultivation of dermatophytes
No. per Pack 100
Packaging Type Tube with Tight Cap
Quantity 100 Pack
Show More Show Less

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.