Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Sabouraud Liquid Broth Prepared Media, Modified

Catalog No. L21014 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L21014 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L21014 Supplier BD Supplier No. 221014
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for the cultivation of yeast and molds

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Broth Type Sabouraud Liquid Broth, Modified (Antibiotic Assay Medium #13)
Cap Type Tight Cap
Certifications/Compliance ISO/DIS, 11014, AOAC, USP
Color Beige
Content And Storage 2 to 25°C
Detects Fungi, Tubercle bacilli
Form Liquid
Format Liquid, Tube
Manufacturing Origin United States
Media Type Sabouraud Liquid Broth
Product Type Broth Tube
Sterility Sterile
Target Organism Class Fungi
Test Type Microbial Cultivation
Formulation Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 20g/L
For Use With (Application) Cultivation of Yeast and molds
No. per Pack 10
Packaging Type Tube with Tight Cap
Quantity 10/Pk.
Show More Show Less

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.