Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Schaedler Broth Prepared Media with Vitamin K1

Catalog No. L21542 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L21542 100 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L21542 Supplier BD Supplier No. 221542
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For cultivation of fastidious aerobic and anaerobic microorganisms

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Broth Type Schaedler Broth with Vitamin K1
Certifications/Compliance CE
Color Beige
Content And Storage 2°C to 8°C
Additive Vitamin K1
Detects Fastidious ,Aerobic and Anaerobic Microorganisms
Form Liquid
Manufacturing Origin United States
Media Type Schaedler Broth
Product Type Broth Tube
Sterility Sterile
Target Organism Class Fastidious Organisms
Formulation Pancreatic digest of Casein 8.2g/L, Peptic digest of Animal Tissue 2.5g/L, Papaic digest of Soybean Meal 1g/L, Dextrose 5.8g/L, Yeast extract 5g/L, Sodium Chloride 1.7g/L, Dipotassium Phosphate 0.8g/L, L-Cystine 0.4g/L, Hemin 0.01g/L, Vitamin K1 0.01g/L,
For Use With (Application) Cultivation of fastidious aerobic and anaerobic microorganisms
No. per Pack 100
Packaging Type Tube with Tight Cap
Quantity 100 Pack
Show More Show Less

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.