Learn More
BD BBL™ Mycoflask™ Selective Seven H11 (Mitchison) Agar Prepared Media Slants
Shop All BD Diagnostic Systems ProductsSpecifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014, BS12, CMPH2, MCM9 |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Format | Mycoflask, Agar Slants |
| Manufacturing Origin | United States |
| Media Type | Selective 7H11 |
| Product Type | Agar Slants |
| Sterility | Sterile |
| Formulation | Pancreatic digest of Casein 1g/L, Monosodium Glutamate 0.5g/L, Sodium Citrate 0.4g/L, Pyridoxine 1mg/L, Biotin 0.5mg/L, Ferric Ammonium Citrate 0.04g/L, Ammonium Sulfate 0.5g/L, Disodium Phosphate 1.5g/L, Monopotassium Phosphate 1.5g/L, Magnesium Sulfate |
| For Use With (Application) | Isolation and cultivation of pathogenic mycobacteria from potentially contaminated specimens |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.