Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Selenite-F Broth Prepared Media

Catalog No. BB21020 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10 Pack
100 Pack
Media Type:
Selenite-F Broth
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Media Type
BB21020 10 Pack Selenite-F Broth
L21021 100 Pack Selenite-F Broth
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BB21020 Supplier BD Supplier No. 221020
Add to Cart
Add to Cart
This item is not returnable. View return policy

For isolation of Salmonella from feces, urine, water, foods and other materials of sanitary importance

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BS12, MCM9, SMWW
Color Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Selenite-F Broth
Formulation Pancreatic digest of Casein 5g/L, Lactose 4g/L, Sodium Selenite 4g/L, Sodium Phosphate 10g/L
For Use With (Application) Isolation of Salmonella from feces, urine, Water, foods and other materials of sanitary importance
Packaging Type Tube with Tight Cap
Quantity 10 Pack

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.