Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ SIM Medium (Sulfide, Indole, Motility)

Catalog No. BB21010 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BB21010 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BB21010 Supplier BD Supplier No. 221010
Add to Cart
Add to Cart

Used for differentiating enteric bacilli on the basis of sulfide production, indole formation and motility

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BAM
Color Yellow Brown
Form Liquid
Manufacturing Origin United States
Media Type SIM Medium (Sulfide, Indole, Motility)
Product Type Agar Tube
Sterility Sterile
Formulation Pancreatic digest of Casein 20g/L, Peptic digest of Animal Tissue 6.1g/L, Ferrous Ammonium Sulfate 0.2g/L, Sodium Thiosulfate 0.2g/L, Agar 3.5g/L
For Use With (Application) Used for differentiate enteric Bacilli on the basis of Sulfide production, indole formation and motility
No. per Pack 10
Packaging Type Tube with Tight Cap
Quantity 10/Pk.
Show More Show Less

For in vitro diagnostic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.