Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Simmons Citrate Agar Prepared Media Slants

Catalog No. BB21026 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BB21026 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BB21026 Supplier BD Supplier No. 221026
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used for differentiation of gram-negative bacteria on the basis of citrate utilization

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Appearance Medium to Dark Green, Slear to Slightly Hazy
Certifications/Compliance CE, AOAC, BAM, CCAM, COMPF, ISO
Color Green
Content And Storage 2°C to 8°C
Additive Bromthymol Blue
Form Liquid
Format Prepared Media Slant, Tube
Manufacturing Origin United States
Media Type Simmons Citrate Agar
Product Type Agar Slants
Sterility Sterile
Target Organism Class Bacteria
Test Type Microbial Differentiation
Formulation Ammonium Dihydrogen Phosphate 1g/L, Dipotassium Phosphate 1g/L, Sodium Chloride 5g/L, Sodium Citrate 2g/L, Magnesium Sulfate 0.2g/L, Agar 15g/L, Bromthymol blue 0.08g/L
For Use With (Application) Isolation and Identification of Microorganisms
Includes 10 Slants, Tight Caps
Incubation Conditions 35 ±2°C; Aerobic Atmosphere
Incubation Time 4 Days
No. per Pack 10
Packaging Type Tube with Tight Cap
Quantity 10/Pk.
Manufacturing Quality AOAC, BAM, CCAM, COMPF, ISO, USDA
Show More Show Less

For in vitro diagnostic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.