Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Thioglycollate (Fluid) Prepared Media Enriched with Vitamin K1 and Hemin

Catalog No. L97642 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
ISO/DIS, 11014, BS12, CMPH2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
L97642 ISO/DIS, 11014, BS12, CMPH2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L97642 Supplier BD Supplier No. 297642
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used for the cultivation of a wide variety of microorganisms, particularly obligate anaerobes

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Broth Type Thioglycollate Medium, Fluid, Enriched with Vitamin K1 and Hemin
Certifications/Compliance ISO/DIS, 11014, BS12, CMPH2
Color Light Yellow
Content And Storage 15°C; to 30°C
Additive Vitamin K1, Hemin, Sodium Thioglycollate
Detects Microorganisms, Tubercle bacilli, Fungi, Anaerobes, Aerobes, Microaerophiles
Form Liquid
Format Prepared Media, Tube
Manufacturing Origin United States
Media Type Thioglycollate Medium
Product Type Prepared Microbiology Tubes
Sterility Sterile
Target Organism Class Fungi
Test Type Microbial Cultivation
Formulation Pancreatic digest of Casein 15g/L, L-Cystine 0.5g/L, Dextrose 5g/L, Yeast extract 5g/L, Sodium Chloride 2.5g/L, Sodium Thioglycollate 0.5g/L, Resazurin 0.001g/L, Agar 0.75g/L, Hemin 0.1g/L, Vitamin K1 2mg/L
For Use With (Application) Cultivation of wide variety of microorganisms, particularly obligate anaerobes
Includes Tube, Tight Cap
Incubation Conditions 30°C to 35°C; Aerobic
Incubation Time 18 to 48 hr. (up to 72 hr.), Up to 5 Days
No. per Pack 100
Packaging Type Tube with Tight Cap
Quantity 100 Pack
Protein Source Casein, Yeast
Source Milk (Casein), Yeast
Manufacturing Quality USP
Show More Show Less

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.