Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Thioglycollate Prepared Media with Calcium Carbonate

Catalog No. L98518 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Content And Storage:
2 to 25°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
L98518 2 to 25°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L98518 Supplier BD Supplier No. 298518
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for the cultivation of a wide variety of microorganisms, particularly obligate anaerobes

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, BS12, CLSI, CMPH2, MCM9
Content And Storage 2 to 25°C
Form Liquid
Manufacturing Origin United States
Media Type Thioglycollate Medium
Sterility Sterile
Formulation Pancreatic digest of Casein 17g/L, Papaic digest of Soybean Meal 3g/L, Dextrose 6g/L, Sodium Chloride 2.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.7g/L, Sodium Sulfite 0.1g/L, 1/ tube Marble Chip
For Use With (Application) Cultivation of anaerobic microorganisms Thioglycollate media
Packaging Type Glass Tube with Cap
Quantity 100 Pack

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.