Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Thioglycollate Prepared Media Enriched with Vitamin K1 and Hemin

Catalog No. L4321788 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10 x 8 mL
100 x 5 mL
100 x 8 mL
100 x 10 mL
Color:
Beige
Light Yellow
Packaging Type:
Size A tube with tight cap
Size K tube with tight cap
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color Packaging Type
L4321788 100 x 8 mL Light Yellow Size K tube with tight cap
L21742 100 x 5 mL Beige Size K tube with tight cap
BB21787 10 x 8 mL Light Yellow Size K tube with tight cap
L97292 100 x 10 mL Beige Size A tube with tight cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. L4321788 Supplier BD Supplier No. 221788
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for use in the isolation and cultivation of fastidious or slow growing, obligately anaerobic microorganisms present in clinical materials

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BS12, CLSI, CMPH2, MCM9
Color Light Yellow
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Thioglycollate Medium
Formulation Pancreatic digest of Casein 17g/L, Papaic digest of Soybean Meal 3g/L, Dextrose 6g/L, Sodium Chloride 2.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.7g/L, L-Cystine 0.25g/L, Sodium Sulfite 0.1g/L, Hemin 0.005g/L, Vitamin K1 0.001g/L
For Use With (Application) Isolation and cultivation of fastidious or slow growing, obligately anaerobic microorganisms present in clinical materials
Packaging Type Size K tube with tight cap
Quantity 100 x 8 mL

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.