Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Thioglycollate Prepared Media without Indicator-135C

Catalog No. p-4638842 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
8 mL
For Use With (Application):
For recovery of wide variety of microorganisms, particularly obligate anaerobes, from clinical specimens and other materials
Packaging Type:
Glass Tube with Cap
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity For Use With (Application) Packaging Type
BB21200 8 mL For recovery of wide variety of microorganisms, particularly obligate anaerobes, from clinical specimens and other materials Glass Tube with Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BB21200 Supplier BD Supplier No. 221200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for the recovery of a wide variety of microorganisms from clinical specimens and other materials

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, BS12, CMPH2
Color Light Yellow
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Thioglycollate Medium
Formulation Pancreatic digest of Casein 17g/L, Papaic digest of Soybean Meal 3g/L, Dextrose 6g/L, Sodium Chloride 2.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.7g/L, L-Cystine 0.25g/L, Sodium Sulfite 0.1g/L
For Use With (Application) For recovery of wide variety of microorganisms, particularly obligate anaerobes, from clinical specimens and other materials
Packaging Type Glass Tube with Cap
Quantity 8 mL

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.