Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Todd Hewitt Broth Prepared Media with Gentamicin and Nalidixic Acid

Catalog No. L99486 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
CE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
L99486 CE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L99486 Supplier BD Supplier No. 299486
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used for the selective enrichment of group B streptococci (Streptococcus agalactiae)

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE
Color Light Yellow
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Todd Hewitt Broth
Formulation Heart infusion from (solids) 3.1g/L, Peptone 20g/L, Dextrose 2g/L, Sodium Chloride 2g/L, Sodium Phosphate 0.4g/L, Sodium Carbonate 2.5g/L, Gentamicin 8mg/L, Nalidixic Acid 15mg/L
For Use With (Application) Selective enrichment of group B streptococci
Packaging Type Tube with Tight Cap
Quantity 100 Pack

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.