Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Todd Hewitt Broth Prepared Media

Catalog No. L21714 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L21714 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L21714 Supplier BD Supplier No. 221714
Add to Cart
Add to Cart

Used for the cultivation of beta-hemolytic streptococci

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Broth Type Todd Hewitt Broth
Cap Type Tight Cap
Certifications/Compliance CE
Content And Storage 2 to 25°C
Detects Tubercle bacilli and FungI
Form Liquid
Format Liquid, Prepared Media, Tube
Manufacturing Origin United States
Media Type Todd Hewitt Broth
Product Type Broth Tube
Sterility Sterile
Target Organism Class Bacteria
Test Type Microbial Cultivation
Fill Volume 5 mL
Formulation Heart infusion from (solids) 3.1g/L, Peptone 20g/L, Dextrose 2g/L, Sodium Chloride 2g/L, Sodium Phosphate 0.4g/L, Sodium Carbonate 2.5g/L
For Use With (Application) Cultivation of beta-hemolytic streptococci
Includes 100 Tubes
No. per Pack 100
Packaging Type Tube with Tight Cap
Quantity 5 mL
Manufacturing Quality CE
Show More Show Less

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.