Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Trypticase™ Soy Agar Slants Prepared Media

Catalog No. BB21087 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10/Pk.
100 Pack
Packaging Type:
Tube
Tube with Tight Cap
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
BB21087 100 Pack Tube with Tight Cap
BB21086 10/Pk. Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BB21087 Supplier BD Supplier No. 221087
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for the isolation and cultivation of fastidious and nonfastidious microorganisms, including anaerobic and aerobic bacteria

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP
Color Beige
Content And Storage 2 to 25°C
Form Liquid
Manufacturing Origin United States
Media Type Trypticase Soy Agar
Formulation Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 15g/L
For Use With (Application) Presumptive identification of Helicobacter pylori in gastric antral biopsy specimens
Packaging Type Tube with Tight Cap
Quantity 100 Pack

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.