Learn More
BD BBL™ Mycoflask™ Trypticase™ Soy Broth Prepare Media with 20% Glycerol
Shop All BD Diagnostic Systems ProductsSpecifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Beige |
| Content And Storage | 2 to 25°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | TSB with 20% Glycerol |
| Formulation | Pancreatic digest of Casein 17g/L, Papaic digest of Soybean Meal 3g/L, Dextrose 2.5g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Glycerol 200mL |
| For Use With (Application) | Long-term frozen maintenance of bacterial stock cultures |
| No. per Pack | 10 |
| Packaging Type | Tube |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.