Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Trypticase™ Soy Broth Prepare Media with 20% Glycerol

Catalog No. p-4639811 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
1.5 mL, 10/Pk.
1.5 mL, 100/Pk.
10 mL, 100/Pk.
Packaging Type:
Ctn. of 100 size K tubes
Tube
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
L97352 10 mL, 100/Pk. Tube
BB97808 1.5 mL, 100/Pk. Ctn. of 100 size K tubes
L96346 1.5 mL, 10/Pk. Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. L97352 Supplier BD Supplier No. 297352
Only null left
Add to Cart
Add to Cart

Used in the long-term frozen maintenance of bacterial stock cultures

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Beige
Content And Storage 2 to 25°C
Form Liquid
Manufacturing Origin United States
Media Type TSB with 20% Glycerol
Formulation Pancreatic digest of Casein 17g/L, Papaic digest of Soybean Meal 3g/L, Dextrose 2.5g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Glycerol 200mL
For Use With (Application) Long-term frozen maintenance of bacterial stock cultures
No. per Pack 100
Packaging Type Tube
Quantity 10 mL, 100/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.