Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Trypticase™ Soy Broth Prepare Media with 6.5% Sodium Chloride

Catalog No. BB21351 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
CE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
BB21351 CE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BB21351 Supplier BD Supplier No. 221351
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for the cultivation of fastidious and nonfastidious microorganisms from a variety of clinical and nonclinical specimens

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE
Color Light Yellow
Form Liquid
Manufacturing Origin United States
Media Type Trypticase Soy Broth with 6.5% Sodium Chloride
Formulation Pancreatic digest of Casein 17g/L, Papaic digest of Soybean Meal 3g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Dextrose 2.5g/L
For Use With (Application) For qualitative procedures for the cultivation of fastidious and non-fastidious microorganisms
Packaging Type Glass Tube
Quantity 100 Pack

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.