Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ TSI Agar™ Slants (Triple Sugar Iron Agar) Prepared Media

Catalog No. BB21039 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BB21039 100/Pk.
BB21038 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BB21039 Supplier BD Supplier No. 221039
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for the cultivation of fastidious and nonfastidious microorganisms from a variety of clinical and nonclinical specimens

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, ISO, MCM9, SMD, SMWW, USDA
Color Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type TSI Agar (Triple Sugar Iron)
Formulation Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Sodium Chloride 5g/L, Lactose 10g/L, Sucrose 10g/L, Dextrose 1g/L, Ferrous Ammonium Sulfate 0.2g/L, Sodium Thiosulfate 0.2g/L, Phenol red 0.025g/L, Agar 13g/L
For Use With (Application) Cultivation of fastidious and non-fastidious microorganisms from variety of clinical and nonclinical specimens
Packaging Type Size K tubes with tight cap
Quantity 100/Pk.

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.