Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Urea Broth (Rapid) Prepared Media

Catalog No. B98330 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
CE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
B98330 CE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B98330 Supplier BD Supplier No. 298330
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for the presumptive identification of Helicobacter pylori in gastric antral biopsy specimens

Vials with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Broth Type Urea Broth (Rapid)
Certifications/Compliance CE
Color Clear
Content And Storage 2°C to 8°C
Form Liquid
Format Liquid
Manufacturing Origin United States
Media Type Urea Broth
Product Type Broth Tube
Sterility Sterile
Fill Volume 0.5 mL
Formulation Pancreatic digest of Gelatin 1g/L, Dextrose 1g/L, Sodium Chloride 5g/L, Potassium Phosphate 2g/L, Urea 20g/L, Phenol red 0.012g/L
For Use With (Application) For presumptive identification of Helicobacter pylori in gastric antral biopsy specimens
No. per Pack 10
Packaging Type Vial
Quantity 0.5 mL
Show More Show Less

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.