Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Urease Test Broth (Stuart's) Prepared Media

Catalog No. L21719 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
3 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L21719 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L21719 Supplier BD Supplier No. 221719
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used for the differentiation of organisms, especially the Enterobacteriaceae, on the basis of urease production

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Broth Type Urease Test Broth
Cap Type Tight Cap
Certifications/Compliance CE, AOAC, BAM, COMPF, SMD
Color Yellow
Content And Storage 2°C to 8°C
Form Liquid
Format Liquid, Tube, Prepared Media
Manufacturing Origin United States
Media Type Urease Test Broth
Product Type Broth Tube
Formulation Urea 200g/L, Monopotassium Phosphate 91g/L, Disodium Phosphate 95g/L, Yeast extract 1g/L, Phenol red 0.1g/L
For Use With (Application) For differentiation of organisms, especially Enterobacteriaceae, on basis of Urease production
Includes 10 Tubes
No. per Pack 10
Packaging Type Tube with Tight Cap
Quantity 3 mL
Show More Show Less

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.