Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Oxacillin Screen Agar (Mueller Hinton Agar w/6μg/mL Oxacillin, 4% NaCl) Prepared Media

Catalog No. B21952X Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Oxacillin screen agar; 10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
B21952X Oxacillin screen agar; 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B21952X Supplier BD Supplier No. 221952
This item is not returnable. View return policy

For the detection of methicillin-resistant Staphylococcus aureus (MRSA)

  • Ready-to-use
  • 100 X 10mm and 100 X 15mm style plates

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Form Liquid
Manufacturing Origin United States
Media Type Oxacillin screen agar (Mueller Hinton agar with 6μg/mL oxacillin and 4% NaCl)
Formulation Beef extract 2g/L, Acid Hydrolysate of Casein 17.5g/L, Starch 1.5g/L, Agar 17g/L, Sodium Chloride 40g/L, Oxacillin 6mg/L
For Use With (Application) Methicillin resistant Staphylococcus aureus
Packaging Type Plate
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.