Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Bottled Media: Buffered Sodium Chloride Peptone Solution (pH 7.0)

Catalog No. B257087 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Certifications/Compliance:
ISO/DIS, 11014
USP, EP, JP
Packaging Type:
Sterile prepared infusion bottle, 10/Pk.
Sterile prepared infusion bottle, 25/Pk
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Certifications/Compliance Packaging Type
B257087 500 mL USP, EP, JP Sterile prepared infusion bottle, 10/Pk.
B257086 100 mL ISO/DIS, 11014 Sterile prepared infusion bottle, 25/Pk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B257087 Supplier BD Supplier No. 257087
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for dissolving, suspending and diluting test samples

  • Used to make suspensions of organisms for testing growth promoting and inhibitory properties of media when examining non-sterile pharmaceutical products for specified microorganisms
  • Fluid provides osmotic stability, stable pH value
  • Maintains viability of microorganisms during preparation of samples
  • Surface-active ingredients or inactivators of antimicrobial agents
TRUSTED_SUSTAINABILITY

Specifications

Appearance Colorless, Clear
ATCC® 16404, 6633, 10231, 8739, 9027, 6538
Broth Type Buffered Sodium Chloride Peptone Solution
Cap Type Stopper with Ring
Certifications/Compliance USP, EP, JP
Color Clear
Content And Storage 5°C to 25°C
Country of Origin United States
Detects Microorganisms
Form Liquid
Format Bottle
Manufacturing Origin United States
Media Type Buffered Sodium Chloride Peptone solution
Preparation Method Ready-to-Use
Product Type Peptone
Sterility Sterile
Target Organism Class Non-fastidious Organisms
Test Type Media Testing
Fill Volume 500 mL
Formulation Proteose Peptone No.3 1,0g/L, Potassium Dihydrogen Phosphate 3.6g/L, Disodium Hydrogen Phosphate (Dihydrate) 7.2g/L, Sodium Chloride 4.3g/L
For Use With (Application) Detection of micororganisms from pharmaceutical, food, environmental or Water testing samples
Includes Injection stopper and pull ring
Incubation Time 2 hr.
Packaging Type Sterile prepared infusion bottle, 10/Pk.
Product Line BBL
Quantity 500 mL
Specimen Type Food, Water
Manufacturing Quality USP, EP, JP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.