Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Media Mossel Enterobacteriaceae Enrichment Broth

Catalog No. B292627 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Content And Storage:
2°C to 8°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
B292627 2°C to 8°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B292627 Supplier BD Supplier No. 292627
Add to Cart
Add to Cart

For isolation of bile-tolerant gram-negative bacteria from nonsterile pharmaceutical products

  • Prepared Bottled Media, ready for immediate use
  • Wide mouth bottle
  • Used for selectively enriching and detecting Enterobacteriaceae in food
  • With transparent labeling to allow clear view of media and culture
  • Meets United States Pharmacopeia (USP), European Pharmacopoeia (EP) and Japanese Pharmacopoeia (JP)1-3 performance specifications
  • Appearance: Medium to dark green and clear to trace hazy
  • Reaction at 25°C, pH: 7.2 ± 0.2
  • Volume: 90mL, Packaging: 10 Each

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, COMPF, EP, ISO, JP, USP
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Mossel Enterobacteriaceae Enrichment Broth
Formulation Pancreatic digest of Gelatin 10g/L, Dextrose 5g/L, Oxgall 20g/L, Disodium Phosphate 8g/L, Monopotassium Phosphate 2g/L, Brilliant green 15mg/L
For Use With (Application) Detection of Enterobacteriaceae, type: Bottle, wide mouth
Packaging Type Tube with Tight Cap
Quantity 90 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.