Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Media Sabouraud Dextrose Agar Deeps

Catalog No. B96182 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B96182 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B96182 Supplier BD Supplier No. 296182
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in qualitative procedures for cultivation of pathogenic and nonpathogenic fungi (particularly dermatophytes)

  • Prepared media with chloramphenicol
  • Pour tubes, Grouping: Q through S products
  • Appearance: Light to medium tan cream to yellow; trace hazy
  • Reaction at 25°C, pH: 5.6 ± 0.2
  • Meets United States Pharmacopeia (USP), European Pharmacopoeia (EP) and Japanese Pharmacopoeia (JP)1-3 performance specifications
  • Volume: 20mL

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BAM, CCAM, CMPH2, COMPF, EP, JP, MCM9, USP
Color Light Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Sabouraud Dextrose Agar
Product Type Agar Tube
Target Organism Class Fungi
Test Type Fungal Testing
Formulation Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 40g/L, Agar 15g/L
For Use With (Application) For qualitative procedures for cultivation of pathogenic and nonpathogenic fungi (particularly dermatophytes)
No. per Pack 100
Packaging Type Pour Tube with Cap
Quantity 20 mL
Protein Source Casein, Animal Tissue
Source Animal
Manufacturing Quality USP, EP, JP
Show More Show Less

For in vitro diagnostic Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.