Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Media Trypticase™ Soy Agar

Catalog No. B240901 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
ISO/DIS, 11014
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
B240901 ISO/DIS, 11014
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B240901 Supplier BD Supplier No. 299623
This item is not returnable. View return policy

Prepared contact plates for environmental monitoring

  • With Lecithin and Polysorbate 80 (double bag)
  • For detection and enumeration of microorganisms present on surfaces of sanitary importance
  • Dehydrated Appearance: Medium fine, softly lumped powder, free of extraneous material
  • Prepared Appearance: Light to medium, yellow to tan, slightly to moderately hazy
  • Reaction of 4.57%, at 25°C, pH: 7.3 ± 0.2

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 2°C to 8°C
Additive Lecithin, Polysorbate 80
Form Liquid
Manufacturing Origin United States
Media Type Trypticase™ Soy Agar with Lecithin and Polysorbate
Sterility Sterile
Fill Volume 500 mL
Formulation Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Lecithin 0.7g/L, Polysorbate 80 5g/L, Agar 15g/L
For Use With (Application) Detection and enumeration of microorganisms present on surfaces of sanitary importance
Packaging Type Double Bag
Quantity 500 mL
Show More Show Less

For Laboratory Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.