Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated: CHROMagar™ Orientation Media

Catalog No. B215081 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100/Pk.
Certifications/Compliance:
ISO/DIS, 11014
Color:
Light Beige
Media Type:
Orientation Agar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Certifications/Compliance Color Media Type
B215081 100/Pk. ISO/DIS, 11014 Light Beige Orientation Agar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B215081 Supplier BD Supplier No. 215081
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nonselective, differential medium for presumptively identifying bacterial isolates from primary clinical specimens

  • Visual differentiationn of colonies is enhanced
  • Allows isolation of both Gram positive and Gram negative pathogens

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Detects Bacterial Isolates, Enterococcus faecalis, Escherichia coli, Proteus mirabilis
Form Liquid
Manufacturing Origin United States
Media Type Orientation Agar
Product Type Prepared Microbiology Plate
Sterility Sterile
Formulation Chromopeptone 16.1g/L, Chromogen Mix 1.3g/L, Agar 15g/L
For Use With (Application) Isolation, differentiation and enumeration of urinary tract pathogens
Incubation Conditions 35 ±2°C, Aerobic
Incubation Time 18 to 24 hr.
Packaging Type Plate
Quantity 100/Pk.
Protein Source Chromopeptone
Manufacturing Quality cGMP Standards, FDA-inspected, ISO 9001 Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.