Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: BiGGY Agar (Nickerson Agar)

Catalog No. L97254 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
BiGGY Agar (Nickerson Agar); 20/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
L97254 BiGGY Agar (Nickerson Agar); 20/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L97254 Supplier BD Supplier No. 297254
This item is not returnable. View return policy

For the isolation of microorganisms from samples or specimens

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Biggy Agar (Nickerson Agar)
Packaging Type Plate
Quantity 20/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.