Learn More
BD BBL™ Prepared Plated Media: Brain Heart Infusion Agar (Deep Fill)
Shop All BD Diagnostic Systems ProductsDescription
- To reduce the effects of drying during prolonged incubation, the plates can be deep filled
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Light Beige |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Brain heart infusion Agar (deep fill) |
| Formulation | Brain heart infusion from (solids) 8g/L, Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Casein 16g/L, Sodium Chloride 5g/L, Dextrose 2g/L, Disodium Phosphate 2.5g/L, Agar 13.5g/L |
| For Use With (Application) | Cultivation of microorganisms |
| Packaging Type | Plate |
| Quantity | 100/Pk. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.