Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Campy CVA Agar

Catalog No. L97246 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
100/Pk.
20/Pk.
Weight (Metric):
1130 g
4000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Weight (Metric)
L97246 20/Pk. 1130 g
B97713 100/Pk. 4000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. L97246 Supplier BD Supplier No. 297246

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective media used in the primary isolation and cultivation of Campylobacter jejuni from human fecal specimens

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Dark Red
Form Liquid
Format Prepared Plate
Manufacturing Origin United States
Media Type Campylobacter Cefoperazone Vancomycin-Amphotericin B Agar
Formulation Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Dextrose 1g/L, Yeast extract 2g/L, Sodium Chloride 5g/L, Sodium Bisulfite 0.1g/L, Agar 15g/L, Cefoperazone 20mg/L, Vancomycin 10mg/L, Amphotericin B 2mg/L, Sheep Blood, defibrinated
For Use With (Application) Campylobacter jejuni from Human feces
Packaging Type Plate
Quantity 20/Pk.
Weight (Metric) 1130 g
Manufacturing Quality cGMP Standards, FDA-inspected, ISO 9001 Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.