Learn More
BD BBL™ Prepared Plated Media: Campylobacter Agar with 5 Antimicrobics and 10% Sheep Blood (Blaser)
Shop All BD Diagnostic Systems ProductsDescription
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Red |
| Content And Storage | 2°C to 8°C |
| Form | Solid |
| Manufacturing Origin | United States |
| Media Type | Campylobacter Agar with 5 antimicrobics and 10% Sheep Blood (Blaser) |
| Formulation | Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Dextrose 1g/L, Yeast extract 2g/L, Sodium Chloride 5g/L, Sodium Bisulfite 0.1g/L, Agar 15g/L, Amphotericin B 2mg/L, Cephalothin 15mg/L, Trimethoprim 5mg/L, Vancomycin 10mg/L, Polymyxin B 250U, Sheep Blood, defibrinated 10% |
| For Use With (Application) | Campylobacter jejuni from Human feces |
| Packaging Type | Plate |
| Quantity | 20/Pk. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.