Learn More
BD BBL™ Prepared Plated Media: Chocolate II Agar (GC II Agar with Hemoglobin and IsoVitaleX™)
Shop All Waters Corporation ProductsDescription
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Dark Brown |
| Content And Storage | 2°C to 8°C |
| Form | Agar |
| Manufacturing Origin | United States |
| Media Type | Chocolate II Agar (GC II Agar with Hemoglobin and IsoVitaleX™) |
| Product Type | Agar Plate |
| Sterility | Sterile |
| Target Organism Class | Fastidious Organisms |
| Test Type | Microbial Cultivation |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.