Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: CHROMagar™ Candida

Catalog No. B4354093 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
ISO/DIS, 11014
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
B4354093 ISO/DIS, 11014
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B4354093 Supplier BD Supplier No. 254093
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For solation and presumptive identification of yeast and filamentous fungi and differentiation of Candida albicans, C. tropicalis and C. krusei

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Beige
Content And Storage 2°C to 8°C
Country of Origin Australia, New Zealand, Canada, United States, China
Additive Chromogen Mix
Form Liquid
Manufacturing Origin United States
Media Type Chromagar™ Candida
Product Type Prepared Microbiology Plate
Sterility Sterile
Target Organism Class Fungi
Test Type Microbial Identification
Formulation Chromopeptone 10g/L, Glucose 20g/L, Chromogen Mix 2g/L, Chloramphenicol 0.5g/L, Agar 15g/L
For Use With (Application) Yeast and filamentous fungi and differentiation of Candida strains
Incubation Conditions 35 ±2°C
Incubation Time 36 to 48 hr.
Packaging Type Plate
Quantity 20/Pk.
Manufacturing Quality ISO 9001:2008, FDA-registered, ISO 13485:2003
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.