Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: CHROMagar™ Staph aureus

Catalog No. 1443241 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
ISO/DIS, 11014
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
14-432-41 ISO/DIS, 11014
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-432-41 Supplier BD Supplier No. 214982
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective medium for the isolation, enumeration and identification of Staphylococcus aureus from clinical and food sources

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Content And Storage 2°C to 8°C
Detects Staphylococcus aureus
Form Liquid
Manufacturing Origin United States
Media Type CHROMagar Staph aureus
Target Organism Class Gram-positive Organisms
Test Type Microbial Identification
Formulation Chromopeptone 40g/L, Sodium Chloride 25g/L, Chromogen Mix 0.5g/L, Inhibitory Agents 0.07g/L, Agar 14g/L
For Use With (Application) Staphylococcus aureus
Incubation Time 24 hr.
Packaging Type Plate
Quantity 20/Pk.
Weight (Metric) 1200 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.