Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: CIN Agar (Yersinia Selective)

Catalog No. l21848 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10/Pk.
For Use With (Application):
Isolation of Y. enterocolitica
Weight (Metric):
800 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity For Use With (Application) Weight (Metric)
L21848 10/Pk. Isolation of Y. enterocolitica 800 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L21848 Supplier BD Supplier No. 221848
This item is not returnable. View return policy

For the isolation of Yersinia enterocolitica from a variety of clinical and nonclinical specimens

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Red
Form Solid
Manufacturing Origin United States
Media Type Cefsulodin Irgasan™ Novobiocin Agar
Formulation Pancreatic digest of Gelatin 10g/L, Peptic digest of Animal Tissue 5g/L, Beef extract 5g/L, Yeast extract 2g/L, Mannitol 20g/L, Sodium Pyruvate 2g/L, Sodium Chloride 1g/L, Magnesium Sulfate 0.001g/L, Sodium Desoxycholate 0.5g/L, Agar 12g/L, Crystal Violet 0.001g/L, Neutral red 0.03g/L, Cefsulodin 15mg/L, Irgasan 4mg/L, Novobiocin 2.5mg/L
For Use With (Application) Isolation of Y. enterocolitica
Packaging Type Plate
Quantity 10/Pk.
Weight (Metric) 800 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.