Learn More
Description
- Ready-to-use
- Non-sterile
- High viscous liquid
- Yellow tint light brown hazy
- 100 X 10mm and 100 X 15mm style plates
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Yellow |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Columbia agar |
| Sterility | Non-sterile |
| Formulation | Pancreatic digest of Casein 14.5g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 14g/L, Growth Factors 1.5g/L, selective Agents 40.2g/L, Sheep Blood, defibrinated 5% |
| For Use With (Application) | Group A streptococci |
| Incubation Time | 18 to 72 hr. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.