Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Columbia Agar with 5% Sheep Blood

Catalog No. b21263x Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21263X 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B21263X Supplier BD Supplier No. 221263
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

For the isolation of microorganisms from samples or specimens

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Red
Form Solid
Manufacturing Origin United States
Media Type Columbia Agar with 5% Sheep Blood
Formulation Pancreatic digest of Casein 12g/L, Peptic digest of Animal Tissue 5g/L, Yeast extract 3.5g/L, Beef extract 3g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Sheep Blood, Defibrinated 5%
For Use With (Application) Non-fastidious and fastidious microorganisms
Packaging Type Plate
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.