Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Corn Meal Agar with Polysorbate 80 (Deep Fill)

Catalog No. b21854x Shop All BD Diagnostic Systems Products
Change view
Click to view available options
:
Corn Meal Agar with Polysorbate 80 (Deep Fill); 10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
B21854X Corn Meal Agar with Polysorbate 80 (Deep Fill); 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B21854X Supplier BD Supplier No. 221854
This item is not returnable. View return policy

For the testing of Candida species for their ability to produce chlamydospores and for the cultivation of fungi

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates
  • Deep-filled plates reduce effects of drying during prolonged incubation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Form Liquid
Manufacturing Origin United States
Media Type Corn Meal Agar with Polysorbate 80
Formulation Corn Meal infusion from (solids) 2g/L, Agar 15g/L, Polysorbate 80 10mL
For Use With (Application) Testing Candida species producing Chlamydospores and general purpose for cultivating fungi
Packaging Type Plate
Quantity 10/Pk.
Weight (Metric) 1300 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.