Learn More
Description
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Light Beige |
| Form | Solid |
| Manufacturing Origin | United States |
| Media Type | Hektoen enteric Agar |
| Formulation | Peptic digest of Animal Tissue 12g/L, Yeast extract 3g/L, Bile Salts 9g/L, Lactose 12g/L, Sucrose 12g/L, Salicin 2g/L, Sodium Chloride 5g/L, Sodium Thiosulfate 5g/L, Ferric Ammonium Citrate 1.5g/L, Bromthymol blue 0.064g/L, Acid Fuchsin 0.1g/L, Agar 13.5g/L |
| For Use With (Application) | Gram negative enteric microorganisms, especially Shigella |
| Packaging Type | Plate |
| Quantity | 20/Pk. |
| Weight (Metric) | 1170 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.